missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Streptavidin HP SpinTrap™ Kit

Product Code. 11540644 Shop All Cytiva Products
Change view
Click to view available options
:
16 spin columns
Product Code.
11540644 16 spin columns
1 options
This item is not returnable. View return policy

Product Code. 11540644

Brand: Cytiva 28903130

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Prepacked single-use spin columns for simple, small-scale protein enrichment through immobilized biotinylated biomolecules

  • Can also be used for the direct enrichment of proteins that are biotinylated
  • Fast and flexible protocol with elution conditions formatted for both electrophoresis and LC-MS analysis workflows
  • Useful for exploiting either the strong interaction of biotin and streptavidin or the somewhat weaker interaction of 2-iminobiotin and streptavidin
  • Each column is packed with Streptavidin Sepharose™ High Performance Medium; for use with a standard microcentrifuge
  • Easy scale-up with HiTrap™ Streptavidin HP prepacked columns

Streptavidin Sepharose HP Medium

  • Matrix: rigid, highly cross-linked 6% agarose
  • Ave. particle size: 34μm
  • Binding capacity: >300nmol biotin/mL medium; 6mg biotinylated BSA/mL medium
  • pH stability: 4 to 9 (long term)
Streptavidin HP SpinTrap Columns
  • Medium volume: 100μL
  • Max. sample loading volume: 600μL

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity >300nM Biotin/mL
Flow Rate <150 cm/hr.
Kit Contents Streptavidin Sepharose HP, spin column and 96-well filter plates filter plates
Material Polypropylene, Polyethylene
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4°C to 8°C
Particle Size 34 μm
Quantity 16 Columns
For Use With (Application) Gel electrophoresis, liquid chromato graphy
Type SpinTrap Kit
compliance-icons
Product Identifier
  • streptavidin HP spintrap
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.