missing translation for 'onlineSavingsMsg'
Learn More

BD Streptavidin HRP Solution

Product Code. 10151040
Click to view available options
Quantity:
1 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10151040

Brand: BD 554066

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Streptavidin is a non-glycosylated protein that is prepared chromatographically from the bacterium Streptomyces avidinii. Streptavidin homotetramers have a particularly high, non-covalent binding affinity for biotin.

  • Each investigator should titrate the reagent to obtain optimal results
  • Use in ELISA, immunocytochemistry, and immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Conjugate HRP
pH 7.2
Quantity 1 mL
Content And Storage Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze.
Form Liquid
For Use With (Application) ELISA
Buffer Aqueous buffered solution containing BSA and ProClin™ 150 as preservative
compliance-icons
Product Identifier
  • Streptavidin-HRP ELISA
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501-Dispose of contents/container to an appropriate treatment and disposal facility in accordancewith applicable laws and regulations, a.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.