missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Streptavidin Mag Sepharose™ Magnetic Bead
Beschreibung
Streptavidin Mag Sepharose uses the strong interaction between biotin and streptavidin ligand, which is immobilized on magnetic beads.
- High-capacity small-scale purification of proteins from cell lysates and biological fluids
- Provides high purity and yield
- Allows easy parallel screening of proteins with high repeatability
- Simple enrichment of proteins from small or large sample volumes (low μl to high ml scale)
- The beads are available in two pack sizes: 2 × 1mL 10% medium slurry and 5 × 1mL 10% medium slurry
- A milliliter of 10% medium slurry is the same as 100 μl sedimented medium which is sufficient for 20 purification runs according to the recommended immunoprecipitation protocol
- Binding capacity: >300μg biotinulated BSA/mL of medium slurry
- Stability: stable
- Immunoprecipitation
- Purification
- Biomarker reseach
Spezifikation
Spezifikation
| Color | White |
| Quantity | 2 x 1 mL |
| Content And Storage | 4°C to 8°C |
| Form | Liquid |
| For Use With (Application) | For simple and efficient enrichment of target Proteins by immunoprecipitation and purification of Biotinylated biomolecules |
Sicherheit und Handhabung
Produktidentifikator
- Streptavidin Mag Sepharose, 2 x 1 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: ethanol
- UNKNOWN ACUTE-66.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 50% of components with unknown hazards to the aquatic environment.
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?