missing translation for 'onlineSavingsMsg'
Learn More

BD Streptavidin, PE-CF594

Product Code. 15801819
Click to view available options
Quantity:
0.1 mg
25 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15801819

Brand: BD 562318

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Developed as a better alternative to PE-Texas Red, for use in Flow Cytometry

The Streptavidin is conjugated to BD Horizon PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496 nm and 564 nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red (eg 610/20-nm filter).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.2 mg/mL
Conjugate PE-CF594
Quantity 25 μg
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.