missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin Protein, DyLight™ 405

Product Code. 10464535
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10464535 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10464535 Supplier Invitrogen™ Supplier No. 21831

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protein

Streptavidin Protein, DyLight 405 conjugate, 21831, from Invitrogen., 21831, from Invitrogen. Species Reactivity: ; Applications: ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot.

Thermo Scientific Streptavidin DyLight Conjugates are preparations of streptavidin biotin-binding protein that are tagged with Thermo Scientific DyLight Fluors for use in high-performance fluorescence assays and cellular imaging. The streptavidin biotin-binding protein is offered as highly functional conjugates with each of nine different DyLight Fluorescent Dyes having absorption spectra ranging from 350nm to 777nm and bright emission spectra that match the principal output wavelengths of common fluorescence microscopes and imagers. DyLight Conjugates exhibit higher fluorescence intensity and photostability than Alexa Fluor*, Cy* and LI-COR* Dyes in many applications and remain highly fluorescent over a broad pH range (pH 4-9).
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
Conjugate DyLight 405
Cross Reactivity Tag
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.2
pH 7.2
Quantity 1 mL
Regulatory Status RUO
Content And Storage 4°C
Form Liquid
For Use With (Application) ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Purity or Quality Grade purified
Host Species N/A
Product Type Streptavidin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.