missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin Protein, DyLight™ 488

Product Code. 10229973
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 10229973

Brand: Invitrogen™ 21832

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protein

Streptavidin Protein, DyLight 488 conjugate, 21832, from Invitrogen., 21832, from Invitrogen. Species Reactivity: ; Applications: ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot.

Thermo Scientific Streptavidin DyLight Conjugates are preparations of streptavidin biotin-binding protein that are tagged with Thermo Scientific DyLight Fluors for use in high-performance fluorescence assays and cellular imaging. The streptavidin biotin-binding protein is offered as highly functional conjugates with each of nine different DyLight Fluorescent Dyes having absorption spectra ranging from 350nm to 777nm and bright emission spectra that match the principal output wavelengths of common fluorescence microscopes and imagers. DyLight Conjugates exhibit higher fluorescence intensity and photostability than Alexa Fluor*, Cy* and LI-COR* Dyes in many applications and remain highly fluorescent over a broad pH range (pH 4-9).
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
Conjugate DyLight 488
Cross Reactivity Tag
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.2
pH 7.2
Quantity 1 mL
Regulatory Status RUO
Content And Storage 4°C
Form Liquid
For Use With (Application) ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Purity or Quality Grade purified
Host Species N/A
Product Type Streptavidin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.