missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptavidin Protein, DyLight™ 594
Description
21842 contains 1 mL of protein in magenta colored liquid at a concentration of 1 mg/mL.
Streptavidin is a tetrameric protein purified from Streptomyces avidinii that binds very tightly to the vitamin biotin with a Kd of ~ 10-14 mol/l. The high affinity recognition of biotin and biotinylated molecules has made streptavidin one of the most important components in diagnostics and laboratory kits.
Specifications
Specifications
| Concentration | 1 mg/mL, 1mg/mL |
| For Use With (Application) | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Formulation | PBS with 1% BSA and 0.02% sodium azide; pH 7.2 |
| Name | Streptavidin |
| Conjugate | DyLight 594 |
| Cross Reactivity | Tag |
| pH Range | 7.2 |
| Purification Method | Purified |
| Quantity | 1 mL |
| Storage Requirements | 4°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?