missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Streptavidin Sepharose™ High Performance Medium

Product Code. 11565015 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11565015 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11565015 Supplier Cytiva Supplier No. 17511301
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Used for convenient, fast, reliable purification of biotinylated compounds. Streptavidin Sepharose™ High Performance Medium is a physically and chemically stable medium.

  • Extremely useful for exploiting either the strong interactions of biotin and streptavidin or the somewhat weaker interactions of 2-iminobiotin and streptavidin
  • Binding capacity of Streptavidin: >300nmol biotin/mL medium; 6mg biotinylated BSA/mL medium
  • Stable (long-term) in the pH range 4 to 9
  • Average particle size: 34μm
  • Ideal for packing into Tricorn™ 5/50 GL, XK 16/20, or other columns where larger-scale purifications are required

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity >300nM Biotin/mL
Flow Rate <150 cm/hr.
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4°C to 8°C
Particle Size 34 μm
Quantity 5 mL
For Use With (Application) Affinity chromatography
Type Agarose
compliance-icons
Product Identifier
  • Streptavidin Sepharose High Performance, 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.