missing translation for 'onlineSavingsMsg'
Learn More

Streptomycin sulfate, MedChemExpress

Product Code. 16488498
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488498 10 g 10g
16404377 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488498 Supplier Medchem Express Supplier No. HYB0472
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Streptomycin sulfate is an aminoglycoside antibiotic, that inhibits protein synthesis.

Streptomycin sulfate is an aminoglycoside antibiotic, that inhibits protein synthesis.

Specifications

Chemical Name or Material Streptomycin sulfate
CAS 3810-74-0
Color White
Purity Grade Notes Research
Molecular Formula C21H42N7O18S1.5
Quantity 10 g
Solubility Information H2O : ≥ 100 mg/mL (137.23 mM) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O[C@@]1(C=O)[C@H]([C@@H](O[C@H]1C)O[C@]([C@@H]([C@@H](O)[C@@H]2NC(N)=N)O)([H])[C@H]([C@@H]2O)NC(N)=N)O[C@@](O[C@@H](CO)[C@H](O)[C@H]3O)([H])[C@H]3NC.[1.5H2SO4]
Molecular Weight (g/mol) 728.69
Formula Weight 728.69
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.