missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Stringknits™ 76-200 Series Lightweight Nylon Gloves

Product Code. 12930135 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
12 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12930135 Medium 12 pairs
12920135 Small 12 pairs
12950135 X-Large 12 pairs
12940135 Large 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12930135 Supplier Ansell™ Supplier No. 76200080
  • ONLINE EXCLUSIVE PRICE
    29.00 €
    19.02 € / 12 pairs
    Save 9.98 €
    34% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Permits longer, more comfortable wear with no weak points which might split or fray. Ansell Stringknits™ 76-200 Series Lightweight Nylon Gloves can be worn on their own or as liners.

  • Good value product protection
  • No seams to rub or irritate
  • Ambidextrous
  • Allows good air flow to the hand, preventing heat and sweat
  • Washable at 40°C without shrinkage or loss of shape
  • Soft, comfortable, flexible and low lint
  • Applications include general handling, delicate manipulation, fine assembly of dry parts
TRUSTED_SUSTAINABILITY

Specifications

Back Material Nylon
Construction Seamless Knit
Cuff Material Nylon
Coating Material Uncoated
Color White
Exterior Finish No Texture
Grade 13 gauge
Hand Ambidextrous
Material Nylon
Product Type Lightweight Gloves
Size Medium
Glove Cuff Color White
Glove Cuff Style Knit
Liner Material Nylon
Lining Lined
Quantity 12 Pairs
Resistant To Mechanical protection
Series 76-200
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.