missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Strip Holder

Brand:  Cytiva 80-6416-87

Code : GE

Additional Details : Weight : 0.17700kg

 View more versions of this product

Product Code. 10036044

  • 532.00 € / Each

Estimated Shipment: 19-05-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Ettan™ IPGphor™ 3 IEF System
6 x Strip holder cover, 1 x 60mL strip holder cleaning solution, instructions
7cm
Strip Holder
6 Pack
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers