missing translation for 'onlineSavingsMsg'
Learn More

Strontium aluminate, 99.5% (metals basis)

Product Code. 11364597
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11364597

Brand: Thermo Scientific Alfa Aesar 039647.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12004-37-4 | SrAl2O4

Strontium aluminate acts as a photo luminescent phosphor with long persistence of phosphorescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12004-37-4
Molecular Formula SrAl2O4
MDL Number MFCD00054054
Synonym strontium aluminate, strontium;oxido oxo alumane, dioxo-2,4-dioxa-1,5-dialumina-3-strontapentane, strontium aluminate,-100 mesh trace metals basis

Specifications

Physical Form Powder
Quantity 100 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Slightly soluble in water.
Formula Weight 205.58
Density 3.559
Chemical Name or Material Strontium aluminate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.