missing translation for 'onlineSavingsMsg'
Learn More

Strontium Chloride, 99-102%, Certified AR for Analysis, Fisher Chemical™

Product Code. 11493723 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493723 500 g 500g
11423673 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493723 Supplier Fisher Chemical Supplier No. S/8320/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10025-70-4
Molecular Formula Cl2H12O6Sr
Molecular Weight (g/mol) 266.61
MDL Number MFCD00149865
InChI Key AMGRXJSJSONEEG-UHFFFAOYSA-L
IUPAC Name strontium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]

Specifications

Melting Point 115°C
Color White
pH 5.0 to 7.0
Physical Form Solid
Quantity 500 g
Formula Weight 266.62g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Strontium chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.