missing translation for 'onlineSavingsMsg'
Learn More

Strontium Chloride Hexahydrate, 99 to 103%, ACS Reagent, Honeywell Fluka™

Product Code. p-7120844 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15622750 100 g Poly Bottle 100g
15632750 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15622750 Supplier Honeywell-Fluka Supplier No. 255521100G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-70-4
Molecular Formula Cl2H12O6Sr
Molecular Weight (g/mol) 266.61
MDL Number MFCD00149865
InChI Key AMGRXJSJSONEEG-UHFFFAOYSA-L
IUPAC Name strontium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]

Specifications

Boiling Point 1250°C
Melting Point 874°C
Physical Form Solid
Quantity 100 g
Linear Formula SrCl2 · 6H2O
UN Number NONH for all modes of transport
Formula Weight 266.62g/mol
Percent Purity 99.0 to 103.0%
Grade ACS Reagent
Packaging Poly Bottle
Density 1.930g/cm3 at 20°C
Chemical Name or Material Strontium Chloride Hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.