missing translation for 'onlineSavingsMsg'
Learn More

Strontium chromate, Alfa Aesar™

Brand:  Alfa Aesar 089026.A1

Code : NEW

Additional Details : CAS Number : 7789-06-2 Weight : 1.00000kg Transport : UN number : 3087 Chem class : 5.1 Pack group : III

Quantity 1kg

 View more versions of this product

Product Code. 15472987

  • 222.00 € / 1kg

Estimated Shipment date
from Supplier 17-09-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

7789-06-2
203.61
NVKTUNLPFJHLCG-UHFFFAOYSA-N
24599
[O-][Cr](=O)(=O)[O-].[Sr+2]
CrO4Sr
MFCD00049549
strontium chromate, strontium yellow, strontium chromate vi, sutokuro t, deep lemon yellow, strontium chromate a, c.i. pigment yellow 32, strontium chromate 1:1, strontium chromate 12170
strontium;dioxido(dioxo)chromium

Specifications

Strontium chromate
CrO4Sr
strontium chromate, strontium yellow, strontium chromate vi, sutokuro t, deep lemon yellow, strontium chromate a, c.i. pigment yellow 32, strontium chromate 1:1, strontium chromate 12170
[O-][Cr](=O)(=O)[O-].[Sr+2]
203.61
203.61
3.895g/mL
7789-06-2
MFCD00049549
NVKTUNLPFJHLCG-UHFFFAOYSA-N
strontium;dioxido(dioxo)chromium
24599
Powder
1kg
Videos
SDS Documents