missing translation for 'onlineSavingsMsg'
Learn More

Strontium hydroxide octahydrate, 98%, pure

Product Code. 11904141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
1kg
5kg
Product Code. Quantity Packaging unitSize
11904141 5 kg Plastic Bottle 5kg
10441671 1 kg Glass bottle 1kg
2 options
This item is not returnable. View return policy

Product Code. 11904141

Brand: Thermo Scientific Acros 270230050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1311-10-0 | H18O10Sr | 265.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1311-10-0
Molecular Formula H18O10Sr
Molecular Weight (g/mol) 265.75
MDL Number MFCD00150395
InChI Key UJPWWRPNIRRCPJ-UHFFFAOYSA-L
IUPAC Name strontium(2+) octahydrate dihydroxide
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Sr++]

Specifications

Melting Point 100.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 kg
Assay Percent Range 98%
Linear Formula Sr(OH)2·8H2O
Merck Index 14,8843
Solubility Information Solubility in water: 7.0g/L (20°C). Other solubilities: soluble in hot water, soluble in acids
Formula Weight 265.75
Percent Purity 98%
Grade Pure
Packaging Plastic Bottle
Chemical Name or Material Strontium hydroxide octahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Strontium hydroxide octahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.