Learn More
Strontium hydroxide octahydrate, tech.
Description
Strontium hydroxide octahydrate is used in the sugar industry for its purification, oil and paint industry to improve its drying properties and as an active ingredient in depilatory hair removal products. It is also used as a raw material for manufacturing of other products of strontium salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100°C-8H2O |
| Physical Form | Crystalline |
| Quantity | 10 kg |
| Solubility Information | Soluble in hot water and acid. Insoluble in acetone. |
| Formula Weight | 265.76 (121.60 Anhydrous) |
| Grade | Technical |
| Sensitivity | Air sensitive; Hygroscopic |
| Density | 1.9 |
| Chemical Name or Material | Strontium hydroxide octahydrate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.