missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium isopropoxide, 96%
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Spezifikation
| Melting Point | 130°C (decomposition) |
| Quantity | 1 g |
| UN Number | UN3205 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Soluble in isoprpanol. |
| IUPAC Name | strontium(2+) bis(propan-2-olate) |
| Formula Weight | 205.82 |
| Percent Purity | 96% |
| Physical Form | Powder |
| Chemical Name or Material | Strontium isopropoxide |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur