missing translation for 'onlineSavingsMsg'
Learn More

Strontium isopropoxide, 96%

Product Code. 11392428
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11392428

Brand: Thermo Scientific Alfa Aesar 014481.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88863-33-6 | C6H14O2Sr | 205.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88863-33-6
Molecular Formula C6H14O2Sr
Molecular Weight (g/mol) 205.80
MDL Number MFCD00050487
InChI Key OHULXNKDWPTSBI-UHFFFAOYSA-N
Synonym strontium isopropoxide, strontium 2+ bis propan-2-olate, strontium propan-2-olate, diisopropoxystrontium, acmc-20egm0, strontium isopropylate, 2-propanol, strontiumsalt 9ci, strontium 2+ ion bis propan-2-olate
PubChem CID 6099237
SMILES [Sr++].CC(C)[O-].CC(C)[O-]

Specifications

Melting Point 130°C (decomposition)
Quantity 25 g
UN Number UN3205
Sensitivity Air and moisture sensitive
Solubility Information Soluble in isoprpanol.
IUPAC Name strontium;propan-2-olate
Formula Weight 205.82
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Strontium isopropoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.