missing translation for 'onlineSavingsMsg'
Learn More

Strontium Nitrate Anhydrous, Certified AR for Analysis, Fisher Chemical

Product Code. 10214450 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10214450 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10214450 Supplier Fisher Chemical Supplier No. S/8402/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 645°C
CAS 10042-76-9
Color White
pH 5.0 to 7.0
Physical Form Crystalline Solid
Quantity 500 g
Molecular Formula N2O6Sr
MDL Number MFCD00011248
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name strontium(2+) dinitrate
Molecular Weight (g/mol) 211.63
PubChem CID 24848
Formula Weight 211.63g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Strontium nitrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.