missing translation for 'onlineSavingsMsg'
Learn More

Strontium nitrate, Puriss., ≥98%, Honeywell Fluka™

Product Code. 15641380 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
50 kg
Unit Size:
1kg
50kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15641380 50 kg 50kg
15621380 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15641380 Supplier Honeywell-Fluka Supplier No. 1391450KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss.,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
PubChem CID 24848
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 570°C (literature)
CAS Max % 100.0000%
Quantity 50 kg
Linear Formula Sr(NO3)2
UN Number UN1507
IUPAC Name strontium(2+) dinitrate
Formula Weight 211.63
Percent Purity >98%
Grade puriss.
Packaging Fiberboard Box
Density 2.980 g/cm3
Chemical Name or Material Strontium nitrate, Puriss., ≥98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.