missing translation for 'onlineSavingsMsg'
Learn More

Strontium nitrate, Puriss. p.a., ACS Reagent, ≥99.0% (complexometric), Honeywell Fluka™

Product Code. 15673480 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Fiberboard Box
Unit Size:
50kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15673480 50 kg Fiberboard Box 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15673480 Supplier Honeywell-Fluka Supplier No. 3163350KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 570°C (literature)
CAS 10042-76-9
CAS Max % 100.0000%
Quantity 50 kg
Molecular Formula N2O6Sr
Linear Formula Na2S2O3 · 5H2O
MDL Number MFCD00011248
UN Number UN1507
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sr+2]
IUPAC Name strontium;dinitrate
Molecular Weight (g/mol) 211.63
PubChem CID 24848
Formula Weight 211.63
Percent Purity >99%
Grade ACS Reagent
Packaging Fiberboard Box
Density 2.980 g/cm3
Assay complexometric
Chemical Name or Material Strontium nitrate, Puriss. p.a., ACS Reagent, ≥99.0% (complexometric)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.