missing translation for 'onlineSavingsMsg'
Learn More

Strontium titanium oxide, 99+% (metals basis)

Product Code. 11351199
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
Product Code. Quantity unitSize
11351199 1 kg 1kg
11341199 250 g 250g
2 options
This item is not returnable. View return policy

Product Code. 11351199

Brand: Thermo Scientific Alfa Aesar 011399.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in electronics and in electrical insulation | CAS: 12060-59-2 | O3SrTi | 183.48 g/mol

Strontium titanium oxide is used as a diamond stimulant and also used as a strontium-90 in radioisotope thermoelectric generators. It is used in the electronics and electrical insulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-59-2
Molecular Formula O3SrTi
Molecular Weight (g/mol) 183.48
MDL Number MFCD00049554
InChI Key VEALVRVVWBQVSL-UHFFFAOYSA-N
Synonym strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
PubChem CID 82899
IUPAC Name strontium;dioxido(oxo)titanium
SMILES [Sr++].[O-][Ti]([O-])=O

Specifications

Melting Point 2060°C
Physical Form Powder
Quantity 1 kg
Solubility Information Insoluble in water and most solvents.
Formula Weight 183.51
Percent Purity ≥99%
Odor Odorless
Density 4.810 g/mL
Assay (metals basis)
Chemical Name or Material Strontium titanium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.