missing translation for 'onlineSavingsMsg'
Learn More

Strontium Titanium Oxide Substrate, 10 x 10 x 0.5mm, Polished Two Sides, 100 Orientation, Thermo Scientific™

Product Code. 11344158
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344158 1 Ea. Each
11354158 5 Ea. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344158 Supplier Thermo Scientific Alfa Aesar Supplier No. 041439.KT
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

100 Orientation

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-59-2
Molecular Formula O3SrTi
Molecular Weight (g/mol) 183.48
MDL Number MFCD00049554
InChI Key VEALVRVVWBQVSL-UHFFFAOYSA-N
Synonym strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
PubChem CID 82899
SMILES [Sr++].[O-][Ti]([O-])=O

Specifications

Color White
Physical Form Substrate
Quantity 1 Ea.
IUPAC Name strontium(2+) oxotitaniumbis(olate)
Odor Odorless
Chemical Name or Material Strontium Titanium Oxide Substrate, Polished Two Sides
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.