missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Stuart™ Borosilicate Glass Reservoir Bottles

Product Code. 12050892
Change view
Click to view available options
Volume (Metric):
1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12050892 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12050892 Supplier Cole-Parmer™ Supplier No. 1009741

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Product Type Reservoir Bottle
Color Amber
Material Borosilicate Glass
Volume (Metric) 1000 mL
For Use With (Equipment) Pressmatic 2000 For Dispenser A45 Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.