missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SU 16f
GREENER_CHOICE

Product Code. 18084512
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18084512 50 mg 50mg
18054322 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18084512 Supplier Tocris Bioscience™ Supplier No. 3304/50
  • ONLINE EXCLUSIVE PRICE
    1300.00 €
    1225.35 € / 50mg
    Save 74.65 €
    5% Off
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective PDGFRβ inhibitor

Potent and selective platelet-derived growth factor receptor β (PDGFRβ) inhibitor (IC50 = 10 nM) that displays > 14-fold, > 229-fold and > 10000-fold selectivity over VEGFR2, FGFR1 and EGFR respectively. Inhibits proliferation of HUVEC and NIH3T3 cells in vitro (IC50 = 0.11 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 251356-45-3
Molecular Formula C24H22N2O3

Specifications

Quantity 50 mg
Formula Weight 386.44
Purity >98%
Molecular Weight (g/mol) 386.451
Chemical Name or Material SU 16f
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.