missing translation for 'onlineSavingsMsg'
Learn More

SU 6668, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18085152
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18085152 10 mg 10mg
18055682 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085152 Supplier Tocris Bioscience Supplier No. 3335/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

PDGFR, VEGFR and FGFR inhibitor

ATP-competitive PDGFR, VEGF and FGFR inhibitor (IC50 values are 0.06, 2.43, 3.04 and > 100 μM at PDGFRβ, VEGFR2, FGFR1 and EGFR respectively). Inhibits proliferation of HUVEC and NIH3T3 cells in vitro (IC50 values are 0.41, 9.3 and 16.5 μM for VEGF, FGF and PDGF-stimulated growth respectively) and induces > 75% growth inhibition against a broad range of tumor types in vivo. Exhibits antiangiogenic, anti-inflammatory, antimetastatic and proapoptotic activity and is orally active.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 252916-29-3
Molecular Formula C18H18N2O3
Molecular Weight (g/mol) 310.353
InChI Key NHFDRBXTEDBWCZ-NTEUORMPSA-N
Synonym 3-2,4-dimethyl-5-e-2-oxo-1h-indol-3-ylidene methyl-1h-pyrrol-3-yl propanoic acid, e-3-2,4-dimethyl-5-2-oxoindolin-3-ylidene methyl-1h-pyrrol-3-yl propanoic acid, tsu 68, tsu-68 su6668,orantinib, 3-2,4-dimethyl-5-3e-2-oxo-1h-indol-3-ylidene methyl-1h-pyrrol-3-yl propanoic acid, 3-2,4-dimethyl-5-e-2-oxo-1,2-dihydro-3h-indol-3-ylidene methyl-1h-pyrrol-3-yl propanoic acid, tsu-68 random configuration, z-3-2,4-dimethyl-5-2-oxoindolin-3-ylidene-methyl-1h-pyrrol-3-yl propanoic acid, 3-2,4-dimethyl-5-2-oxoindolin-3-ylidene methyl-1h-pyrrol-3-yl propanoic acid, 5-1,2-dihydro-2-oxo-3h-indol-3-ylidene methyl-2,4-dimethyl-1h-pyrrole-3-propanoic acid
PubChem CID 5995546
IUPAC Name 3-[2,4-dimethyl-5-[(E)-(2-oxo-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl]propanoic acid
SMILES CC1=C(NC(=C1CCC(=O)O)C)C=C2C3=CC=CC=C3NC2=O

Specifications

Quantity 10 mg
Formula Weight 310.35
Percent Purity >99%
Chemical Name or Material SU 6668
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.