missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ SUBHT High Throughput Submarine Gel Unit
Description
Includes
- Buffer chamber assembly
- Safety lid, high-voltage leads
- Fluorescent Labeled Running Tray with Casting Gates
- Four 1mm thick 30-well combs
- Colored Loading Strips
- Two Buffer Recirculation Ports
- Fast, error-free loading: load an entire 96-well plate using a multichannel pipetter
- Simplified Analysis: Fluorescent labels for each sample printed on a UV transparent gel tray allows for instant sample identification (matched to a 96-well plate) during post-run visualization
- High Throughput: Run up to 120 samples (96-well plate plus 24 control and/or marker samples) in 30 minutes
- Better Reproducibility: Eliminate variability caused by running buffer pH drift and ionic gradient formation with optional buffer recirculation capability
- Reduced Buffer Needs: Compact tank design needs less buffer to cover gel which not only reduces buffer consumption but provides greater control of voltage gradients and run times
- Maximum Flexibility: Fully compatible with a wide range of combs and accessories to meet all your high throughput and standard gel analysis needs
- This high throughput horizontal gel unit can accommodate up to 120 samples and resolve DNA strands from 50-1000 base pairs in 30 minutes
- Gel Dimensions: 15 x 15 cm
- Maximum Buffer Volume: 1.2L
- Maximum Power Settings: 300V, 200mA, 60W
- Maximum Sample Capacity: 120
- Maximum Running Temperature: 45°C
- Indoor Use: 4-40°C
- Humidity: up to 80%
- Unit Dimensions: 33.5L x 21.5W x 7 cmH
- Safety Certifications: EN61010-1, CE
Applications:
Single Strand Polymorphism (SSP) analysis
Spécification
Spécification
| For Use With (Equipment) | SUBHT High Throughput Submarine Gel Unit |
| Type | Lid |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?