missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Substrate Reagent Pack

Product Code. 18784494
Change view
Click to view available options
Quantity:
16 x 12 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18784494 16 x 12 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18784494 Supplier R&D Systems™ Supplier No. DY999
  • ONLINE EXCLUSIVE PRICE
    374.45 €
    332.85 € / 1 sets
    Save 41.60 €
    11% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

8 vials Color A, 8 vials Color B Product in process of being discontinued and replaced by new Next generation pack

Warm to room temperature before use. Color Reagent A and Color Reagent B should be mixed together in equal volumes within 15 minutes of use. Protect from light. Add 100μL of the mixture to each well and incubate at room temperature for 20 minutes while protecting from light. Add Stop Solution (Catalog # DY994) and read the microplate.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 16 x 12 mL
For Use With (Application) ELISA
Product Line R&D Systems
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.