missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, ACS, 99.0% min

Product Code. p-7050011
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11317588

Brand: Thermo Scientific Alfa Aesar 033272.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-15-6 | C4H6O4 | 118.09 g/mol

Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry; as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

Melting Point 186°C to 188°C
Boiling Point 235°C
Beilstein 1754069
Merck Index 14,8869
Solubility Information Soluble in water (increasing with temperature); Soluble in ethanol,ethyl ether,acetone and methanol; Insoluble in toluene,benzene,carbon disulfide,carbon tetrachloride and petroleum ether.
Formula Weight 118.09
Percent Purity ≤99.0%
Grade ACS
Density 1.56 g/mL
Quantity 500 g
Flash Point 206°C (402°F)
Physical Form Crystalline
Chemical Name or Material Succinic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.