missing translation for 'onlineSavingsMsg'
Learn More

Succinic Acid, BAKER ANALYZED™ ACS Reagent, J.T. Baker™

Product Code. 15577824 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15577824 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15577824 Supplier Avantor J.T.Baker Supplier No. 034605

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use

Specifications

CAS 110-15-6
Melting Point 185.0°C to 191.0°C
Molecular Formula C4H6O4
MDL Number MFCD00002789
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILES OC(=O)CCC(O)=O
IUPAC Name butanedioic acid
Molecular Weight (g/mol) 118.09
PubChem CID 1110
ChEBI CHEBI:15741
Formula Weight 118.09
Grade Meets ACS reagent chemical requirements
Quantity 2.5 kg
Packaging Poly Bottle
Physical Form Powder
Specific Gravity 1L = 1.57kg
Chemical Name or Material Succinic Acid
Show More Show Less
compliance-icons
Product Identifier
  • Succinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Succinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.