missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, disodium salt, 99%, anhydrous

Product Code. 10491821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10491821 100 g Plastic bottle 100g
10636442 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10491821 Supplier Thermo Scientific Acros Supplier No. 208741000
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 150-90-3 | C4H4Na2O4 | 162.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-90-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
MDL Number MFCD00002790
InChI Key ZDQYSKICYIVCPN-UHFFFAOYSA-L
Synonym disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277
PubChem CID 9020
ChEBI CHEBI:63675
IUPAC Name disodium;butanedioate
SMILES C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >300.0°C
Color White
Loss on Drying 0.3% max. (120°C)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Plastic bottle
Linear Formula NaO2CCH2CH2CO2Na
Quantity 100 g
Beilstein 02, 606
Merck Index 15, 8808
Solubility Information Solubility in water: 300g/L (25°C).
Formula Weight 162.05
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material Succinic acid, disodium salt, 0.99
Show More Show Less
compliance-icons
Product Identifier
  • Succinic acid, disodium salt, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.