missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid disodium salt, hexahydrate, >98%, MP Biomedicals™

Product Code. 11466332 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466332 100 g 100g
11476332 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466332 Supplier MP Biomedicals Supplier No. 0210292980
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 150-90-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
InChI Key ZDQYSKICYIVCPN-UHFFFAOYSA-L
Synonym disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277
PubChem CID 9020
ChEBI CHEBI:63675
IUPAC Name disodium;butanedioate
SMILES C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 100 g
Formula Weight 270.1
Percent Purity >98%
Physical Form Powder
Chemical Name or Material Succinic acid disodium salt, hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.