missing translation for 'onlineSavingsMsg'
Learn More

Succinic Acid, Extra Pure, SLR, Fisher Chemical

Product Code. 10714581 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10714581 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10714581 Supplier Fisher Chemical Supplier No. S/8520/53
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Specifications

CAS 110-15-6
Melting Point 185°C
Color White
pH 2.7
Boiling Point 235°C
Packaging Plastic powder jar
Molecular Formula C4H6O4
MDL Number MFCD00002789
Quantity 500 g
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILES OC(=O)CCC(O)=O
IUPAC Name butanedioic acid
Molecular Weight (g/mol) 118.09
PubChem CID 1110
ChEBI CHEBI:15741
Formula Weight 118.09g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.