missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, Free acid, ≥99%, Ultrapure

Product Code. 15875188 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging Pakningsstørrelse
15875188 500 g Plastic bottle 500g
15855338 1 kg Plastic bottle 1kg
15865338 5 kg Plastic bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15875188 Leverandør Thermo Scientific Leverandørnr. J21900.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 110-15-6 | C4H6O4 | 118.088 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.088
MDL Number MFCD0000278
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES C(CC(=O)O)C(=O)O

Specifications

Melting Point 186°C to 188°C
Boiling Point 235°C
Density 1.56
Quantity 500 g
Flash Point >110°C
Packaging Plastic bottle
Physical Form Crystalline Powder
Specific Gravity 1.56
Linear Formula HOOCCH2CH2COOH
Beilstein 1754069
Formula Weight 118.09
Percent Purity ≥99%
Chemical Name or Material Succinic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.