missing translation for 'onlineSavingsMsg'
Learn More

Succinic Acid, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15567824 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15567824 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15567824 Supplier Avantor J.T.Baker Supplier No. 034601

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 110-15-6
Molecular Formula C4H6O4
MDL Number MFCD00002789
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILES OC(=O)CCC(O)=O
IUPAC Name butanedioic acid
Molecular Weight (g/mol) 118.09
PubChem CID 1110
ChEBI CHEBI:15741
Formula Weight 118.09
Grade BAKER ANALYZED™ ACS Reagent
Quantity 500 g
Packaging Poly Bottle
Physical Form Powder
Specific Gravity 1L = 1.57kg
Chemical Name or Material Succinic Acid
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.