missing translation for 'onlineSavingsMsg'
Learn More

Succinic Acid, TRC

Product Code. 30006450
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
100g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006450 100 g 100g
30004221 50 g 50g
30004077 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006450 Supplier Toronto Research Chemicals Supplier No. TRCS688765100G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4 H6 O4
Molecular Weight (g/mol) 118.09
Synonym Succinic Acid, Butanedioic acid, Succinic acid (8CI), 1,2-Ethanedicarboxylic acid, 1,4-Butanedioic acid, A 12084, Amber acid, Asuccin, Biosuccinium, Biosuccinium SA, Dihydrofumaric acid, Katasuccin, NSC 106449, NSC 25949, S 3674, Wormwood acid, Yantar-antitox
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(=O)O

Specifications

InChI Formula InChI=1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)
Formula Weight 118.0266
Quantity 100 g
Chemical Name or Material Succinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.