missing translation for 'onlineSavingsMsg'
Learn More

Succinic Anhydride, 99%

Product Code. 10417000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
50g
5g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10417000 1 kg Plastic bottle 1kg
10744353 5 g Glass Bottle 5g
10639803 50 g Plastic bottle 50g
10786874 250 g Plastic bottle 250g
11906171 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10417000 Supplier Thermo Scientific Acros Supplier No. 158760010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Succinic anhydride, 99%, C4H4O3, CAS Number-108-30-5, 2,5-diketotetrahydrofuran, 2,5-furandione, dihydro, butanedioic anhydride, dihydro-2,5-furandione, dihydrofuran-2,5-dione, rikacid sa, succinic acid anhydride, succinic anhydride, succinyl anhydride, succinyl oxide, 1kg, 17,407, 261.0 deg.C | CAS: 108-30-5 | C4H4O3 | 100.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-30-5
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.07
MDL Number MFCD00005525
InChI Key RINCXYDBBGOEEQ-UHFFFAOYSA-N
Synonym succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa
PubChem CID 7922
ChEBI CHEBI:36595
IUPAC Name oxolane-2,5-dione
SMILES O=C1CCC(=O)O1

Specifications

Melting Point 116.0°C to 120.0°C
Quantity 1 kg
Packaging Plastic bottle
Assay Percent Range 99%
Viscosity 3 mPa.s (125°C)
Beilstein 17,407
Fieser 04,468
Merck Index 15,9
Boiling Point 261.0°C
Solubility Information Solubility in water: 62g/L (20°C). Other solubilities: soluble in chloroform,carbon tetrachloride and,alcohol,slightly soluble in ether,20g/L benzene (20°C)
Formula Weight 100.07
Percent Purity 99%
Chemical Name or Material Succinic anhydride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Succinic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.