missing translation for 'onlineSavingsMsg'
Learn More

Succinimide, 98%

Product Code. 10337371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
500g
5g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10337371 100 g Glass bottle 100g
10757582 5 g Glass bottle 5g
10738162 500 g Glass bottle 500g
13488518 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. Afficher la politique du retour.
Code produit 10337371 Fournisseur Thermo Scientific Acros Code fournisseur 348651000
ChemsChrom Mega Deals
Use promo code 27993
Afficher toutes les promotions
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 123-56-8 | C4H5NO2 | 99.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 123-56-8
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.08
MDL Number MFCD00005495
InChI Key KZNICNPSHKQLFF-UHFFFAOYSA-N
Synonym succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione
PubChem CID 11439
ChEBI CHEBI:9307
IUPAC Name pyrrolidine-2,5-dione
SMILES C1CC(=O)NC1=O

Spécification

Melting Point 121.0°C to 126.0°C
Color Brown to White
Boiling Point 285.0°C to 290.0°C
Flash Point 201°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Beilstein 21,369
Merck Index 15,9001
Solubility Information Solubility in water: 240g/L (20°C). Other solubilities: 1g/24 mL alcohol - 1g/5 mL alcohol (60°C), insoluble in ether and chloroform
Formula Weight 99.08
Percent Purity 98%
Physical Form Powder or Flakes
Chemical Name or Material Succinimide, 98%
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Succinimide
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.