missing translation for 'onlineSavingsMsg'
Learn More

Succinimide, 98%

Product Code. 10757582 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
500g
5g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10757582 5 g Glass bottle 5g
10337371 100 g Glass bottle 100g
10738162 500 g Glass bottle 500g
13488518 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10757582 Supplier Thermo Scientific Acros Supplier No. 348650050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-56-8 | C4H5NO2 | 99.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-56-8
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.08
MDL Number MFCD00005495
InChI Key KZNICNPSHKQLFF-UHFFFAOYSA-N
Synonym succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione
PubChem CID 11439
ChEBI CHEBI:9307
IUPAC Name pyrrolidine-2,5-dione
SMILES C1CC(=O)NC1=O

Specifications

Melting Point 121.0°C to 126.0°C
Color Brown to White
Boiling Point 285.0°C to 290.0°C
Flash Point 201°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 21,369
Merck Index 15,9001
Solubility Information Solubility in water: 240g/L (20°C). Other solubilities: 1g/24 mL alcohol - 1g/5 mL alcohol (60°C), insoluble in ether and chloroform
Formula Weight 99.08
Percent Purity 98%
Physical Form Powder or Flakes
Chemical Name or Material Succinimide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Succinimide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.