missing translation for 'onlineSavingsMsg'
Learn More

Succinimide, 98+%

Product Code. 11425603
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425603 250 g 250g
11415603 1000 g 1000g
11435603 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425603 Supplier Thermo Scientific Alfa Aesar Supplier No. A13503.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 123-56-8 | C4H5NO2 | 99.089 g/mol

Succinimide is used in organic syntheses and in industrial silver plating processes. It is also used as anticonvulsant drugs such as ethosuximide, phensuximide and methsuximide. Further, it is used to form covalent bonds between proteins, peptides and plastics. In addition to this, it is used as an important raw material for the synthesis of heterocyclic compounds through N-acyliminium cyclizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinimide is used in organic syntheses and in industrial silver plating processes. It is also used as anticonvulsant drugs such as ethosuximide, phensuximide and methsuximide. Further, it is used to form covalent bonds between proteins, peptides and plastics. In addition to this, it is used as an important raw material for the synthesis of heterocyclic compounds through N-acyliminium cyclizations.

Solubility
Soluble in water and ethanol. Insoluble in ether and chloroform.

Notes
Incompatible with strong oxidizing agents, acids, bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-56-8
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.089
MDL Number MFCD00005495
InChI Key KZNICNPSHKQLFF-UHFFFAOYSA-N
Synonym succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione
PubChem CID 11439
ChEBI CHEBI:9307
IUPAC Name pyrrolidine-2,5-dione
SMILES C1CC(=O)NC1=O

Specifications

Melting Point 121°C to 126°C
Density 1.41
pH 4 to 6
Boiling Point 287°C to 288°C
Flash Point 201°C (393°F)
Odor Odorless
Quantity 250 g
Beilstein 108440
Merck Index 14,8871
Solubility Information Soluble in water and ethanol. Insoluble in ether and chloroform.
Formula Weight 99.09
Percent Purity ≥98%
Chemical Name or Material Succinimide
Show More Show Less
compliance-icons
Product Identifier
  • Succinimide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Succinimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.