missing translation for 'onlineSavingsMsg'
Learn More

Succinimide, TRC

Product Code. 30133113
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133113 5 g 5g
30006140 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133113 Supplier Toronto Research Chemicals Supplier No. TRCS6894005G
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-56-8
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.09
Synonym 2,5-Diketopyrrolidine, 2,5-Dioxopyrrolidine, Butanimide, Lubrizol 6406, NSC 11204, NSC 13114, NSC 49152, Succinic Acid Imide, Succinic Imide
IUPAC Name pyrrolidine-2,5-dione
SMILES O=C1CCC(=O)N1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H5NO2/c6-3-1-2-4(7)5-3/h1-2H2,(H,5,6,7)
Formula Weight 99.032
Chemical Name or Material Succinimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.