missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Succinimidyl 3-(2-Pyridyldithio)Propionate (SPDP)

Product Code. 11530666
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11530666 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11530666 Supplier Invitrogen™ Supplier No. S1531
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The succinimidyl ester chemically reacts with primary amines and after mild reduction of the dithiol, it can react form a disulfide with other thiols.

Crosslinking reagents are useful for probing the spatial relationships and interactions within and between biomolecules.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable By Thiols
Description SPDP
Molecular Weight (g/mol) 312.36
PEGylated No
Spacer Arm Length 6.8 Å
Content And Storage Upon receipt store at -20°C protected from light.
Cell Permeability Yes
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety NHS Ester, Pyridyldisulfide
Spacer Short (<10 Å)
Form Solid
Quantity 100 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.