missing translation for 'onlineSavingsMsg'
Learn More

Succinimidyl-6-(biotinamido) Hexanoate, TRC

Product Code. 30426513
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426513 1 g 1g
30415463 100 mg -
30418436 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426513 Supplier Toronto Research Chemicals Supplier No. TRCS6902501G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72040-63-2
Molecular Formula C20 H30 N4 O6 S
Molecular Weight (g/mol) 454.54
Synonym Enzotin, EZ-Link NHS-LC-Biotin, N-[5-(Succinimidyloxycarbonyl)pentyl]biotin Amide, N-Hydroxysuccinimidyl 6-(Biotinamido)hexanoate, N-Hydroxysuccinimidyl Biotinamidocaproate, B 1582, NHS-LC-Biotin, B 1582, 6-[(Biotinyl)amino]hexanoic Acid N-Hydroxysuccinimide Ester, 5-(N-Succinimidyloxycarbonyl)pentyl d-Biotinamide
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]hexanoate
SMILES O=C(CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)NCCCCCC(=O)ON3C(=O)CCC3=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H30N4O6S/c25-15(7-4-3-6-14-19-13(12-31-14)22-20(29)23-19)21-11-5-1-2-8-18(28)30-24-16(26)9-10-17(24)27/h13-14,19H,1-12H2,(H,21,25)(H2,22,23,29)/t13-,14-,19-/m0/s1
Formula Weight 454.1886
Chemical Name or Material Succinimidyl-6-(biotinamido) Hexanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.