missing translation for 'onlineSavingsMsg'
Learn More

Succinyl chloride, ca. 95%, AcroSeal™

Product Code. 10588865 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 10588865

Brand: Thermo Scientific Acros 430801000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 543-20-4 | C4H4Cl2O2 | 154.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 190.0°C
Melting Point 16.0°C to 17.0°C
CAS 543-20-4
Color Colorless to Brown
Physical Form Liquid
Quantity 100 mL
Assay Percent Range ca. 95%
Molecular Formula C4H4Cl2O2
Linear Formula ClCOCH2CH2COCl
MDL Number MFCD00000749
Flash Point 76°C
Merck Index 15,9004
Synonym succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate
Solubility Information Solubility in water: decomposes
InChI Key IRXBNHGNHKNOJI-UHFFFAOYSA-N
SMILES C(CC(=O)Cl)C(=O)Cl
IUPAC Name butanedioyl dichloride
Molecular Weight (g/mol) 154.98
PubChem CID 10970
Formula Weight 154.98
Percent Purity ∼95%
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4700 to 1.4720
Specific Gravity 1.407
Density 1.4070g/mL
Chemical Name or Material Succinyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • Succinyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.