missing translation for 'onlineSavingsMsg'
Learn More

Sucralose, Thermo Scientific™

Product Code. 17750689 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17750689 250 g 250g
17740689 100 g 100g
17730689 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17750689 Supplier Thermo Scientific Acros Supplier No. 470092500
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

Alpha Vector SUCRALOSE
Chemical Name or Material Sucralose
CAS 56038-13-2
Melting Point 130°C
Color White
Infrared Spectrum Conforms
Packaging Plastic bottle
Molecular Formula C12 H19 Cl3 O8
MDL Number 03648615
Quantity 250 g
Merck Index XIII 8965
Specific Rotation Condition (20°C, 589 nm) (c=1% aq. soln.)
Specific Rotation 84° to 88°
Synonym Trichlorosucrose
Molecular Weight (g/mol) 397.63
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.