missing translation for 'onlineSavingsMsg'
Learn More

Sucrose, 99% max., Molecular biology reagent grade, MP Biomedicals™

Product Code. 11456491 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456491 500 g 500g
11466491 1 kg 1kg
11476491 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456491 Supplier MP Biomedicals Supplier No. 0219401890
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Color White
Density 1.587g/cm3 at 25°C
Assay Percent Range >99%
Quantity 500 g
Formula Weight 342.3
Grade Molecular Biology Reagent
Physical Form Crystalline Powder
Chemical Name or Material Sucrose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.