missing translation for 'onlineSavingsMsg'
Learn More

Sucrose ACS MP Biomedicals

Product Code. 11417902 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417902 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11417902 Supplier MP Biomedicals Supplier No. 0215258491
  • ONLINE EXCLUSIVE PRICE
    138.00 €
    130.20 € / 1kg
    Save 7.80 €
    5% Off
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity ACS grade sucrose suitable for density gradients, enzymatic assays, and cell culture media.

Specifications

CAS 57-50-1
Density 1.587 at 25 °C (water = 1)
Molecular Formula C12H22O11
MDL Number MFCD00006626
Quantity 1 kg
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 342.30
PubChem CID 5988
ChEBI CHEBI:17992
Formula Weight 342.3
Grade ACS
Chemical Name or Material Sucrose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.