missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose Gel-Loading Dye for DNA Gels, 5X (Contains 40% Sucrose/Mol. Biol.), Fisher BioReagents
Description
This dye includes sucrose instead of glycerol. Add the dye to DNA samples prior to electrophoresis on agarose gels.
Specifications
Specifications
| Absorbance | 0.30 to 0.45 AU at 252nm, 0.47 to 0.62 AU, 0.48 to 0.65 AU |
| Chemical Name or Material | Sucrose Gel-Loading Dye for DNA Gels |
| Name Note | 5X, Contains 40% Sucrose |
| CAS | 57-50-1, 7732-18-5 |
| Grade | Electrophoresis |
| Packaging | Poly Bottle |
| Color | Purple |
| Quantity | 5 mL |
| Physical Form | Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?