missing translation for 'onlineSavingsMsg'
Learn More

Sucrose, MP Biomedicals™

Product Code. 11441732 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441732 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11441732 Supplier MP Biomedicals Supplier No. 04821721
  • ONLINE EXCLUSIVE PRICE
    374.00 €
    353.85 € / 5kg
    Save 20.15 €
    5% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sucrose Ultra Pure, ≥99%, White Crystals

  • It is used as a source of carbon in media and animal diet.
  • 8.5% (w/v) sucrose was used in dielectrophoretic field-flow fractionation (dFFF) that allows the rapid and convenient analysis of bulk cell suspensions.
  • Sucrose is a non-reducing disaccharide composed of glucose and fructose linked via their anomeric carbons.
  • It is obtained commercially from sugarcane, sugar beet, and other plants and used extensively as a food and a sweetener.
  • The mixture of glucose and fructose is known as invert sugar.

Specifications

CAS 57-50-1
Melting Point 177°C to 187°C
Color White
Density 1.587 at 25°C (water = 1)
Molecular Formula C12H22O11
MDL Number MFCD00006626
Quantity 5 kg
Beilstein 90825
Synonym Beet sugar, Cane sugar, b-D-Fructofuranosyl-a-D-glucopyranoside, D-(+)-Saccharose
Solubility Information Soluble in water (1g/0.5 mL at 25°C; 1g/0.2 mL boiling water), ethanol (1g/170 mL) or methanol (1g/100 mL); moderately soluble inglycerol or pyridine.
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Optical Rotation 66.0°±5 (H2O, NH4OH)
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 342.30
Grade ≥99%
Physical Form Crystals
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.